missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-3-octanol, 97+%, ACROS Organics™

Brand:  Acros Organics 379065000

Code : 29

Additional Details : CAS Number : 78-69-3 Weight : 0.46400kg


Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10678044

  • £76.10 / 500mL


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

3, 7-Dimethyl-3-octanol
97.0
≥97%
C10H22O
158.28
78-69-3
100.0
97+%
(CH3)2CH(CH2)3C(CH3)(OH)CH2CH3
MFCD00004482

Specifications

3, 7-Dimethyl-3-octanol
76°C
Clear colorless
78-69-3
Authentic
Glass bottle
Liquid
97+%
(CH3)2CH(CH2)3C(CH3)(OH)CH2CH3
MFCD00004482
0.8280g/mL
193.0°C
500mL
97.0
100.0
1.4310 to 1.4350
≥97%
C10H22O
158.28
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,7-Dimethyl-3-octanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS