missing translation for 'onlineSavingsMsg'
Learn More

3,6-Thioxanthenediamine-10,10-dioxide 97%, ACROS Organics™

Brand:  Acros Organics 420910010

Code : 29

Additional Details : CAS Number : 10215-25-5 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10683732

  • £31.90 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

3, 6-Thioxanthenediamine-10, 10-dioxide
10215-25-5
100.0
97%
260.32
97%
96.0
≥96.0%
C13H12N2O2S
MFCD00041841

Specifications

3, 6-Thioxanthenediamine-10, 10-dioxide
10215-25-5
100.0
Powder
97%
Glass bottle
MFCD00041841
214° to 219°C
97%
96.0
Authentic
≥96.0%
C13H12N2O2S
260.32
Beige to yellow-green
1g
Videos
SDS