missing translation for 'onlineSavingsMsg'
Learn More

3,6-Thioxanthenediamine-10,10-dioxide, 97%, Thermo Scientific Chemicals

Catalog Number p-199794
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10215-25-5
C13H12N2O2S
260.32
MFCD00041841
UPVRZVIJGVFROW-UHFFFAOYSA-N
82456
10,10-dioxo-9H-thioxanthene-3,6-diamine
C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N
This item is not returnable. View return policy
10215-25-5
C13H12N2O2S
260.32
MFCD00041841
UPVRZVIJGVFROW-UHFFFAOYSA-N
82456
10,10-dioxo-9H-thioxanthene-3,6-diamine
C1C2=C(C=C(C=C2)N)S(=O)(=O)C3=C1C=CC(=C3)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 213°C to 215°C
Color Beige to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 9h-thioxanthene-3,6-diamine 10,10-dioxide, 9h-thioxanthene-3,6-diamine, 10,10-dioxide, 3,6-diamino-9h-thioxanthene 10,10-dioxide, 3,6-thioxanthenediamine-10,10-dioxide, 3,7-thiaxanthenediamine-5,5-dioxide, 9h-thioxanthene-3,7-diamine, 5,5-dioxide, 9h-thioxanthene-3,6-diamine,10,10-dioxide, 3,7-diamino-10h-dibenzo b,e thiin-5,5-dione, 3,6-diamino-9h-10??-thioxanthene-10,10-dione, 3,6-diamino-9h-10
Formula Weight 260.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 6-Thioxanthenediamine-10, 10-dioxide, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'