missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[(6-Methylpyrazin-2-yl)oxy]aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-6-methylpyrazin-2-yl oxy aniline, benzenamine, 3-6-methyl-2-pyrazinyl oxy, 3-6-methylpyrazin-2-yloxy phenylamine, benzenamine,3-6-methyl-2-pyrazinyl oxy |
Formula Weight | 201.23 |
Percent Purity | 97% |
Chemical Name or Material | 3-[(6-Methylpyrazin-2-yl)oxy]aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'