missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-fluorobenzyl alcohol, 97%, Thermo Scientific™

Catalog Number p-7031101
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
916420-68-3
C7H5Cl2FO
195.01
MFCD09025388
VGTKYVUXLJVEPF-UHFFFAOYSA-N
3,6-dichloro-2-fluorobenzyl alcohol, 3,6-dichloro-2-fluorophenyl methanol, 3,6-dichloro-2-fluorophenyl methan-1-ol
46737554
(3,6-dichloro-2-fluorophenyl)methanol
OCC1=C(Cl)C=CC(Cl)=C1F
This item is not returnable. View return policy
916420-68-3
C7H5Cl2FO
195.01
MFCD09025388
VGTKYVUXLJVEPF-UHFFFAOYSA-N
3,6-dichloro-2-fluorobenzyl alcohol, 3,6-dichloro-2-fluorophenyl methanol, 3,6-dichloro-2-fluorophenyl methan-1-ol
46737554
(3,6-dichloro-2-fluorophenyl)methanol
OCC1=C(Cl)C=CC(Cl)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 72°C
Quantity 1 g
Formula Weight 195.02
Percent Purity 97%
Chemical Name or Material 3,6-Dichloro-2-fluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'