Learn More
Thermo Scientific Maybridge 3,6-Dibromopyridazine, 5g, 97%
Brand: Thermo Scientific Maybridge MO07136DE
3,6-Dibromopyridazine, 5g, 97%
Code : 31
Additional Details : CAS Number : 17973-86-3 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III