missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromopyrazine-2-carboxamide, 97%, Thermo Scientific™

Catalog Number p-7055312
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1301613-77-3
C5H3Br2N3O
280.907
TWCACYWIPUIXAR-UHFFFAOYSA-N
58269823
3,6-dibromopyrazine-2-carboxamide
C1=C(N=C(C(=N1)Br)C(=O)N)Br
This item is not returnable. View return policy
1301613-77-3
C5H3Br2N3O
280.907
TWCACYWIPUIXAR-UHFFFAOYSA-N
58269823
3,6-dibromopyrazine-2-carboxamide
C1=C(N=C(C(=N1)Br)C(=O)N)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 280.9
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,6-Dibromopyrazine-2-carboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'