missing translation for 'onlineSavingsMsg'
Learn More

3,5-Pyridinedicarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-4509903
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
499-81-0
C7H5NO4
167.12
MFCD00006393
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
OC(=O)C1=CC(=CN=C1)C(O)=O
This item is not returnable. View return policy
499-81-0
C7H5NO4
167.12
MFCD00006393
MPFLRYZEEAQMLQ-UHFFFAOYSA-N
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f
10366
CHEBI:46875
pyridine-3,5-dicarboxylic acid
OC(=O)C1=CC(=CN=C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 22,16
Solubility Information Solubility in water: insoluble
Formula Weight 167.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3, 5-Pyridinedicarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Pyridinedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'