missing translation for 'onlineSavingsMsg'
Learn More

3(5)-Phenyl-1H-pyrazole, 99+%, Thermo Scientific Chemicals

Catalog Number p-2700272
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2458-26-6
C9H8N2
144.18
MFCD00159654
OEDUIFSDODUDRK-UHFFFAOYSA-N
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
17155
5-phenyl-1H-pyrazole
N1N=CC=C1C1=CC=CC=C1
This item is not returnable. View return policy
2458-26-6
C9H8N2
144.18
MFCD00159654
OEDUIFSDODUDRK-UHFFFAOYSA-N
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
17155
5-phenyl-1H-pyrazole
N1N=CC=C1C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 75.0°C to 77.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 23,177
Solubility Information Solubility in water: .. Other solubilities: halogenated solvents,alcohols
Formula Weight 144.18
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 3(5)-Phenyl-1H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 3(5)-Phenyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'