missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(5-Methyl-1H-pyrazol-4-yl)propylamine, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 139.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-(5-Methyl-1H-pyrazol-4-yl)propylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'