missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride, Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzene-1-sulfonyl chloride |
Formula Weight | 258.68 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzenesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'