missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3.5 M Potassium Chloride Electrode Filling Solution (Electrolyte), Reagecon
Optimal electrolyte and filling solution for potentiometric titrations, direct pH measurement, redox of ion measurement where silver/silver chloride or calomel reference electrodes are used.
Brand: Reagecon EFS351
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Electrode reference standard, electrode reference material.
- For potentiometric titrations, direct pH measurement, redox or ion measurement where silver/silver chloride or calomel reference electrodes are used.
- Saturated KCl is only sparingly soluble below 20°C, so if measurements below this temperature are to be carried out, 3.5 M Potassium Chloride remains in solution down to 15°C.
- High quality product: equitransference of the cation/anion combination is as close as possible to being equimobile, constant chloride activity, high electrical conductance, as non-chemically reactive as possible.
Specifications
3.5 mol/L | |
pH electrode filling solution | |
100 mL |
pH Electrode | |
Dropper Bottle |