missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dinitro-N-[4-(trifluoromethyl)phenyl]benzamide, 97%

Brand:  Alfa Aesar™ H59647.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405166

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Dinitro-N-[4-(trifluoromethyl)phenyl]benzamide
355.23
MFCD04509248

Specifications

3,5-Dinitro-N-[4-(trifluoromethyl)phenyl]benzamide
Solid
355.23
1g
97%
MFCD04509248
Videos
SDS