3,5-Dimethylthiophenol, 98%, ACROS Organics™


Overview
£11.96 valid until 30/06/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 259230025

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 38360-81-5 Weight: 0.05255kg Transport: UN number : 2810 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 2.5mL

Product Code. 10053330

Quantity Price
1 £ 29.9 / 2.50mL
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3, 5-Dimethylthiophenol
Name Note 98%
CAS 38360-81-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10S
Linear Formula CH3(C6H3)CH3SH
Formula Weight 138.23
MDL Number MFCD00040226

Specification

Assay ≥97.5%
Physical Form Liquid
Formula Weight 138.23
Quantity 2.5mL
Color Clear yellow
CAS Min % 97.5
EINECS Number 253-901-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3(C6H3)CH3SH
Infrared Spectrum Authentic
CAS 38360-81-5
MDL Number MFCD00040226
Assay Percent Range 98%
Refractive Index 1.5665 to 1.5685 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Packaging Glass bottle
Molecular Formula C8H10S
Density 1.02g/mL
Name Note 98%
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
TSCA TSCA
Chemical Name or Material 3, 5-Dimethylthiophenol
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Special Offers