missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 399330050

Code : 29

Additional Details : CAS Number : 857530-80-4 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 10346404

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
C11H19BN2O2
MFCD05663852
97%
222.09

Specifications

3, 5-Dimethylpyrazole-4-boronic acid pinacol ester
5g
Glass bottle
97%
222.09
177.0° to 180.0°C
857530-80-4
97%
C11H19BN2O2
MFCD05663852
Videos
SDS