missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylpyrazin-2-amine, ≥97%, Thermo Scientific™

Catalog Number p-3485408
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
91678-81-8
C6H9N3
123.159
MFCD00107172
GZZRWBRYYZGTNW-UHFFFAOYSA-N
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
2774764
3,5-dimethylpyrazin-2-amine
CC1=CN=C(C(=N1)C)N
This item is not returnable. View return policy
91678-81-8
C6H9N3
123.159
MFCD00107172
GZZRWBRYYZGTNW-UHFFFAOYSA-N
2-amino-3,5-dimethylpyrazine, 3,5-dimethyl-2-pyrazinamine, 3,5-dimethylpyrazine-2-ylamine, pyrazinamine, 3,5-dimethyl, 2-pyrazinamine, 3,5-dimethyl, 3,5-dimethyl-pyrazinamine, 2-amino-3,5-dimethyl-1,4-diazine, 3,5-dimethylpyrazin-2-ylamine
2774764
3,5-dimethylpyrazin-2-amine
CC1=CN=C(C(=N1)C)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethylpyrazin-2-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'