missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isothiocyanate, 99%, Thermo Scientific Chemicals

Catalog Number p-7081360
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
40046-30-8
C9H9NS
163.238
MFCD00041082
DSMXCADWIFIJEX-UHFFFAOYSA-N
3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
142406
1-isothiocyanato-3,5-dimethylbenzene
CC1=CC(=CC(=C1)N=C=S)C
This item is not returnable. View return policy
40046-30-8
C9H9NS
163.238
MFCD00041082
DSMXCADWIFIJEX-UHFFFAOYSA-N
3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
142406
1-isothiocyanato-3,5-dimethylbenzene
CC1=CC(=CC(=C1)N=C=S)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN2922
Beilstein 6856812
Formula Weight 163.24
Percent Purity 99%
Quantity 5 g
Chemical Name or Material 3,5-Dimethylphenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'