missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole, 99%, Thermo Scientific Chemicals

Catalog Number p-7081989
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
300-87-8
C5H7NO
97.117
MFCD00003156
FICAQKBMCKEFDI-UHFFFAOYSA-N
3,5-dimethylisoxazole, isoxazole, 3,5-dimethyl, 3,5-dimethylisooxazole, 3,5-dwumetyloizoksazolu, 3,5-dwumetyloizoksazolu polish, 3,5-dimethylisoxazol, pubchem8641, isoxazole,5-dimethyl, 3,5-dimethyl-isoxazole
9312
3,5-dimethyl-1,2-oxazole
CC1=CC(=NO1)C
This item is not returnable. View return policy
300-87-8
C5H7NO
97.117
MFCD00003156
FICAQKBMCKEFDI-UHFFFAOYSA-N
3,5-dimethylisoxazole, isoxazole, 3,5-dimethyl, 3,5-dimethylisooxazole, 3,5-dwumetyloizoksazolu, 3,5-dwumetyloizoksazolu polish, 3,5-dimethylisoxazol, pubchem8641, isoxazole,5-dimethyl, 3,5-dimethyl-isoxazole
9312
3,5-dimethyl-1,2-oxazole
CC1=CC(=NO1)C

The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -14°C
Density 0.992
Boiling Point 142°C to 144°C
Flash Point 31°C (87°F)
Refractive Index 1.4415
Quantity 25 g
UN Number UN1993
Beilstein 106324
Solubility Information Insoluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylisoxazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'