missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Formula Weight | 141.13 |
Chemical Name or Material | 3, 5-Dimethylisoxazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'