missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4509416
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
2510-36-3
C6H7NO3
141.126
IJEUISLJVBUNRE-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
75636
3,5-dimethyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(=NO1)C)C(=O)O
This item is not returnable. View return policy
2510-36-3
C6H7NO3
141.126
IJEUISLJVBUNRE-UHFFFAOYSA-N
3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
75636
3,5-dimethyl-1,2-oxazole-4-carboxylic acid
CC1=C(C(=NO1)C)C(=O)O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 141.13
Chemical Name or Material 3, 5-Dimethylisoxazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'