missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzyl bromide, 98%, ACROS Organics™

Brand:  Acros Organics 355561000

Code : 29

Additional Details : CAS Number : 27129-86-8 Weight : 0.15000kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10752125

  • £254.40 / 100g


Call for latest availability
+44 (0)1509 555 500
Add to basket


Specifications

Chemical Identifiers

3, 5-Dimethylbenzyl bromide
97.5
≥97.5%
C9H11Br
MFCD00013539
27129-86-8
100.0
98%
199.09

Specifications

3, 5-Dimethylbenzyl bromide
Light yellow
100g
97.5
Glass bottle
≥97.5%
C9H11Br
MFCD00013539
Authentic
36° to 40°C
27129-86-8
100.0
Low Melting Crystalline Mass
98%
199.09
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Dimethylbenzyl bromide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS