missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-p-anisic acid, 98%, Thermo Scientific™

Catalog Number p-115535
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21553-46-8
C10H12O3
180.2
MFCD00020309
WXVQURJGDUNJCS-UHFFFAOYSA-N
3,5-dimethyl-4-methoxybenzoic acid, 3,5-dimethyl-p-anisic acid, 4-methoxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl anisic acid, benzoic acid, 4-methoxy-3,5-dimethyl, 4-methoxy-3,5-dimethylbenzoicacid, acmc-209flo, ksc496g8f, 3,5-dimethyl-4-methoxy-benzoic acid, 4-methoxy-3,5-dimethyl benzoic acid
88944
4-methoxy-3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1OC)C)C(=O)O
This item is not returnable. View return policy
21553-46-8
C10H12O3
180.2
MFCD00020309
WXVQURJGDUNJCS-UHFFFAOYSA-N
3,5-dimethyl-4-methoxybenzoic acid, 3,5-dimethyl-p-anisic acid, 4-methoxy-3,5-dimethyl-benzoic acid, 3,5-dimethyl anisic acid, benzoic acid, 4-methoxy-3,5-dimethyl, 4-methoxy-3,5-dimethylbenzoicacid, acmc-209flo, ksc496g8f, 3,5-dimethyl-4-methoxy-benzoic acid, 4-methoxy-3,5-dimethyl benzoic acid
88944
4-methoxy-3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1OC)C)C(=O)O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C to 194°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H2(CH3)2CO2H
Quantity 1 g
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3, 5-Dimethyl-p-anisic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethyl-p-anisic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'