missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrrole-2-carbaldehyde, ≥95%, Thermo Scientific™

Catalog Number p-2699529
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2199-58-8
C7H9NO
123.155
MFCD00111522
RDFZYUOHJBXMJA-UHFFFAOYSA-N
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole
270465
3,5-dimethyl-1H-pyrrole-2-carbaldehyde
CC1=CC(=C(N1)C=O)C
This item is not returnable. View return policy
2199-58-8
C7H9NO
123.155
MFCD00111522
RDFZYUOHJBXMJA-UHFFFAOYSA-N
3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole
270465
3,5-dimethyl-1H-pyrrole-2-carbaldehyde
CC1=CC(=C(N1)C=O)C
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula Yes
Quantity 250 mg
Formula Weight 123.15
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethyl-1H-pyrrole-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'