missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%, Thermo Scientific™

Catalog Number p-119649
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22042-79-1
C12H12N2O
200.24
MFCD00051396
VOMRTQQGXWPTJK-UHFFFAOYSA-N
89179
3,5-dimethyl-1-phenylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C2=CC=CC=C2)C)C=O
This item is not returnable. View return policy
22042-79-1
C12H12N2O
200.24
MFCD00051396
VOMRTQQGXWPTJK-UHFFFAOYSA-N
89179
3,5-dimethyl-1-phenylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C2=CC=CC=C2)C)C=O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 128°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl, 3,5-dimethyl-1-phenyl-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,3,5-dimethyl-1-phenyl, 3,5-dimethyl-4-formyl-1-phenylpyrazole, 3,5-dimethyl-1-phenyl-4-pyrazolecarboxaldehyde, 3?5-dimethyl-1-phenylpyrazole-4-carboxaldehyde, 3,5-dimethyl-1-phenyl 1h-pyrazole-4-carbaldehyde, 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbaldehyde #
Formula Weight 200.24
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde, 99%
compliance-icons
Product Identifier
  • 3,5-Dimethyl-1-phenylpyrazole-4-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'