missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dimethoxyphenyl trifluoromethanesulfonate, 97%

Brand:  Alfa Aesar™ H55637.03

Code : NEW

Additional Details : CAS Number : 60319-09-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15451948

  • £15.26 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Dimethoxyphenyl trifluoromethanesulfonate
60319-09-7
286.22
MFCD04039976

Specifications

3,5-Dimethoxyphenyl trifluoromethanesulfonate
Colorless
60319-09-7
Liquid
286.22
1.415g/mL
1g
1.4640
97%
MFCD04039976
Videos
SDS