missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzylamine, 98%, ACROS Organics™

Catalog Number p-4258968
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
34967-24-3
C9H13NO2
167.208
YGZJTYCCONJJGZ-UHFFFAOYSA-N
3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
420973
(3,5-dimethoxyphenyl)methanamine
COC1=CC(=CC(=C1)CN)OC
This item is not returnable. View return policy
34967-24-3
C9H13NO2
167.208
YGZJTYCCONJJGZ-UHFFFAOYSA-N
3,5-dimethoxybenzylamine, 3,5-dimethoxybenzyl amine, 3,5-dimethoxyphenyl methanamine, 1-3,5-dimethoxyphenyl methanamine, 3,5-dimethoxy benzyl amine, 3,5-dimethoxyphenyl methylamine, benzenemethanamine, 3,5-dimethoxy, pubchem7393, 3,5-dimethoxy benzylamine, 3,5-dimethoxy-benzylamine
420973
(3,5-dimethoxyphenyl)methanamine
COC1=CC(=CC(=C1)CN)OC
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White
Density 1.11g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 167.21
Percent Purity ≥97.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3, 5-Dimethoxybenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Dimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'