missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65805.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409916

  • £49.82 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
311.38
MFCD00977943

Specifications

3,5-Dimethoxy-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
Solid
311.38
1g
95%
MFCD00977943
Videos
SDS