missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-hydroxybenzaldehyde, 98%, ACROS Organics™

£13.60 - £104.10

Chemical Identifiers

CAS 134-96-3
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00006943
InChI Key KCDXJAYRVLXPFO-UHFFFAOYSA-N
Synonym syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
PubChem CID 8655
ChEBI CHEBI:67380
IUPAC Name 4-hydroxy-3,5-dimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1O)OC)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10386730 SDS View Documents Acros Organics
132880050
5g Glass bottle £13.60
5g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
10031390 SDS View Documents Acros Organics
132880250
25g Glass bottle £28.70
25g View alternative products
Save Up to 
»
EU Stock 10 Please to purchase this item. Need a web account? Register with us today!
10066183 SDS View Documents Acros Organics
132881000
100g Glass bottle £104.10
100g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

134-96-3
182.18
KCDXJAYRVLXPFO-UHFFFAOYSA-N
8655
4-hydroxy-3,5-dimethoxybenzaldehyde
C9H10O4
MFCD00006943
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
CHEBI:67380
COC1=CC(=CC(=C1O)OC)C=O

Specifications

3,5-Dimethoxy-4-hydroxybenzaldehyde
Authentic
1.013
Brown to Green-Yellow
5g
97.5
97.5% min. (GC)
MFCD00006943
syringaldehyde, 3,5-dimethoxy-4-hydroxybenzaldehyde, syringic aldehyde, syringylaldehyde, syringealdehyde, benzaldehyde, 4-hydroxy-3,5-dimethoxy, gallaldehyde 3,5-dimethyl ether, 3,5-dimethoxy-4-hydroxybenzene carbonal, 4-hydroxy-3,5-dimethoxy-benzaldehyde, springaldehyde
KCDXJAYRVLXPFO-UHFFFAOYSA-N
4-hydroxy-3,5-dimethoxybenzaldehyde
8655
182.18
98%
1.0130g/mL
Glass bottle
192.0°C to 193.0°C (14.0mmHg)
110.0°C to 113.0°C
134-96-3
100.0
C9H10O4
15, 9145
Solubility in water: insoluble. Other solubilities: very sparingly soluble in petr ether, soluble in alcohol, ether, chloroform, hot benzene, glacial acetic acid
COC1=CC(=CC(=C1O)OC)C=O
182.18
CHEBI:67380
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Dimethoxy-4-hydroxybenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.