missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
Solubility
Soluble in 50 mg/mL, 95% Ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
Solubility
Soluble in 50 mg/mL, 95% Ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∼237°C (decomposition) |
Quantity | 100 g |
Beilstein | 2207864 |
Solubility Information | Soluble in 50mg/mL,95% Ethanol. |
Formula Weight | 154.12 |
Percent Purity | 98% |
Chemical Name or Material | 3,5-Dihydroxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'