missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3771031
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
26153-38-8
C7H6O3
138.12
MFCD00016611
HAQLHRYUDBKTJG-UHFFFAOYSA-N
benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
94365
CHEBI:50204
3,5-dihydroxybenzaldehyde
OC1=CC(C=O)=CC(O)=C1
This item is not returnable. View return policy
26153-38-8
C7H6O3
138.12
MFCD00016611
HAQLHRYUDBKTJG-UHFFFAOYSA-N
benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
94365
CHEBI:50204
3,5-dihydroxybenzaldehyde
OC1=CC(C=O)=CC(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 153.0°C to 158.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 1 g
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dihydroxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'