missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylhydrazine hydrochloride, 97+%, ACROS Organics™

£25.87 - £73.25

Chemical Identifiers

CAS 134993-88-7
Molecular Formula C6H6F2N2·HCl
Molecular Weight (g/mol) 180.58
MDL Number MFCD03094171
InChI Key CKCNKYAHVKNKHQ-UHFFFAOYSA-N
Synonym 3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
PubChem CID 16217172
IUPAC Name (3,5-difluorophenyl)hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1F)F)NN.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10061114
SDS View Documents
Acros Organics
398830010
1g Glass bottle £25.87
1g View alternative products
Save Up to 
»
Estimated Shipment
11-11-2020
Please to purchase this item. Need a web account? Register with us today!
10346634
SDS View Documents
Acros Organics
398830050
5g Glass bottle £73.25
5g View alternative products
Save Up to 
»
Estimated Shipment
09-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

134993-88-7
180.58
CKCNKYAHVKNKHQ-UHFFFAOYSA-N
16217172
C1=C(C=C(C=C1F)F)NN.Cl
C6H6F2N2·HCl
MFCD03094171
3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
(3,5-difluorophenyl)hydrazine;hydrochloride

Specifications

3, 5-Difluorophenylhydrazine hydrochloride
261.0°C to 266.0°C
134993-88-7
100.0
C6H6F2N2·HCl
MFCD03094171
CKCNKYAHVKNKHQ-UHFFFAOYSA-N
(3,5-difluorophenyl)hydrazine;hydrochloride
16217172
97+%
Glass bottle
1g
97.0
97% min. (HPLC)
F2C6H3NHNH2·HCl
3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
C1=C(C=C(C=C1F)F)NN.Cl
180.58
180.58
Safety and Handling

Safety and Handling

Product Identifier
  • 3,5-Difluorophenylhydrazine hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .