missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenylhydrazine hydrochloride, 97+%, Thermo Scientific™

Catalog Number 10346634
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
134993-88-7
C6H6F2N2·HCl
180.58
MFCD03094171
CKCNKYAHVKNKHQ-UHFFFAOYSA-N
3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
16217172
(3,5-difluorophenyl)hydrazine;hydrochloride
C1=C(C=C(C=C1F)F)NN.Cl
This item is not returnable. View return policy
134993-88-7
C6H6F2N2·HCl
180.58
MFCD03094171
CKCNKYAHVKNKHQ-UHFFFAOYSA-N
3,5-difluorophenylhydrazine hydrochloride, 3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluorophenylhydrazine hcl, 3,5-difluorophenylhydrazinehydrochloride, 1-3,5-difluorophenyl hydrazine hydrochloride, 3,5-difluoro phenylhydrazine, hydrazine, 3,5-difluorophenyl-, monohydrochloride, pubchem1085, pubchem20776, c6h6f2n2.hcl
16217172
(3,5-difluorophenyl)hydrazine;hydrochloride
C1=C(C=C(C=C1F)F)NN.Cl
CAS Min % 97.0
CAS Max % 100.0
Melting Point 261.0°C to 266.0°C
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula F2C6H3NHNH2·HCl
Quantity 5 g
Formula Weight 180.58
Percent Purity 97+%
Chemical Name or Material 3, 5-Difluorophenylhydrazine hydrochloride
compliance-icons
Product Identifier
  • 3,5-Difluorophenylhydrazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'