missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenyl isocyanate, 97%, ACROS Organics™

Brand:  Acros Organics 428230050

Code : 29

Additional Details : CAS Number : 83594-83-6 Weight : 0.06000kg Transport : UN number : 2478 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10067659

  • £107.40 / 5g


Estimated Shipment
15-03-2018
Add to basket


Specifications

Chemical Identifiers

3, 5-Difluorophenyl isocyanate
96.0
97%
F2C6H3NCO
MFCD02093693
83594-83-6
100.0
C7H3F2NO
155.1

Specifications

3, 5-Difluorophenyl isocyanate
54°C
5g
96.0
Glass bottle
97%
F2C6H3NCO
MFCD02093693
1.3100g/mL
159.0°C
83594-83-6
100.0
97%
C7H3F2NO
155.1
Videos
SDS