missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorophenol, 99%, ACROS Organics™

Brand:  Acros Organics 306610010

Code : 29

Additional Details : CAS Number : 2713-34-0 Weight : 0.05100kg Transport : UN number : 2925 Chem class : 1b Pack group : III

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10025963

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3, 5-Difluorophenol
2713-34-0
100.0
99%
F2C6H3OH
MFCD00002255
99%
98.5
99%
C6H4F2O
130.09

Specifications

3, 5-Difluorophenol
White to beige
1g
99%
98.5
Glass bottle
99%
C6H4F2O
130.09
70°C
51.0° to 55.0°C
Authentic
2713-34-0
100.0
Crystals
99%
F2C6H3OH
MFCD00002255
Videos
SDS