missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Description
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.
Solubilit
It is a green alternative to Tetrahydrofuran. is capable of producing high concentrations of bromo, benzyl and allyl Grignard reagents. 2-MeTHF quickly produces clean water phase splits without forming emulsions.
Solubilit
Specifications
CAS | 1417985-27-3 |
Molecular Formula | C7H5BrF2Mg |
MDL Number | MFCD11113509 |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Synonym | 3,5-difluorobenzylmagnesium bromide, 0.25m in 2-methf, qpuypkpzikvhdr-uhfffaoysa-m, 3,5-difluorobenzylmagnesium bromide, 3,5-difluorobenzyl magnesium bromide, grignard reagent |
Solubility Information | Reacts with water. |
InChI Key | QAXANYLHEOPEGM-UHFFFAOYSA-M |
SMILES | [CH2-]C1=CC(=CC(=C1)F)F.[Mg+2].[Br-] |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'