missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-110576
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32085-88-4
C7H4F2O
142.1
MFCD00010329
ASOFZHSTJHGQDT-UHFFFAOYSA-N
benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone
588160
3,5-difluorobenzaldehyde
C1=C(C=C(C=C1F)F)C=O
This item is not returnable. View return policy
32085-88-4
C7H4F2O
142.1
MFCD00010329
ASOFZHSTJHGQDT-UHFFFAOYSA-N
benzaldehyde, 3,5-difluoro, 3,5-difluoro-benzaldehyde, pubchem1414, 3,5difluorobenzaldehyde, acmc-1aewe, 3,5-diflourobenzaldehyde, 3-5-difluorobenzaldehyde, 3, 5-difluorobenzaldehyde, ksc224o7p, 3,5-difluorophenyl methanone
588160
3,5-difluorobenzaldehyde
C1=C(C=C(C=C1F)F)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Green
Density 1.2960g/mL
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CHO
Refractive Index 1.4930 to 1.4950
Quantity 1 g
Specific Gravity 1.296
Formula Weight 142.1
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 3, 5-Difluorobenzaldehyde, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Difluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'