missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroaniline, 98%, ACROS Organics™

Brand:  Acros Organics 311420500

Code : 29

Additional Details : CAS Number : 372-39-4 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10510034

  • £120.80 / 50g


Estimated Shipment
17-05-2018
Add to basket

Specifications

Chemical Identifiers

3, 5-Difluoroaniline
97.5
≥97.5%
C6H5F2N
129.11
372-39-4
100.0
98%
F2C6H3NH2
MFCD00007763

Specifications

3, 5-Difluoroaniline
White to yellow
50g
97.5
Authentic
Product darkens on storage
≥97.5%
C6H5F2N
129.11
75g/mL
37° to 41°C
372-39-4
100.0
Glass bottle
Low Melting Solid
98%
F2C6H3NH2
MFCD00007763
Videos
SDS