missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N,N-diisopropylbenzamide, 97%

Brand:  Alfa Aesar™ H61277.03

Code : NEW

Additional Details : CAS Number : 1288986-11-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447788

  • £131.44 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N,N-diisopropylbenzamide
241.28
21344237
1288986-11-7
MFCD27665223

Specifications

3,5-Difluoro-N,N-diisopropylbenzamide
1288986-11-7
97%
MFCD27665223
1g
Solid
241.28
21344237
Videos
SDS