missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(4-fluoro-2-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62191.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427146

  • £73.56 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(4-fluoro-2-methylphenyl)benzenesulfonamide
301.28
MFCD17629481

Specifications

3,5-Difluoro-N-(4-fluoro-2-methylphenyl)benzenesulfonamide
Solid
301.28
1g
97%
MFCD17629481
Videos
SDS