missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(3-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61955.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408548

  • £79.71 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(3-methylphenyl)benzamide
247.24
MFCD07261386

Specifications

3,5-Difluoro-N-(3-methylphenyl)benzamide
Solid
247.24
1g
97%
MFCD07261386
Videos
SDS