missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(3-fluorobenzyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62596.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447616

  • £79.71 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(3-fluorobenzyl)benzenesulfonamide
C13H10F3NO2S
301.28
MFCD24849049

Specifications

3,5-Difluoro-N-(3-fluorobenzyl)benzenesulfonamide
97%
301.28
1g
C13H10F3NO2S
MFCD24849049
Videos
SDS