missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(3-fluorobenzyl)benzamide, 97%

Brand:  Alfa Aesar™ H61746.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458308

  • £63.71 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(3-fluorobenzyl)benzamide
265.23
MFCD24849011

Specifications

3,5-Difluoro-N-(3-fluorobenzyl)benzamide
Solid
265.23
1g
97%
MFCD24849011
Videos
SDS