missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62618.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497636

  • £63.71 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
337.27
MFCD26143333

Specifications

3,5-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
Solid
337.27
1g
97%
MFCD26143333
Videos
SDS