missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(2-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62433.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407436

  • £63.71 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(2-fluorophenyl)benzenesulfonamide
287.26
MFCD08867833

Specifications

3,5-Difluoro-N-(2-fluorophenyl)benzenesulfonamide
Solid
287.26
1g
97%
MFCD08867833
Videos
SDS