missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(2,5-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61184.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15467688

  • £59.78 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(2,5-difluorophenyl)benzamide
269.2
MFCD04806864

Specifications

3,5-Difluoro-N-(2,5-difluorophenyl)benzamide
Solid
269.2
1g
97%
MFCD04806864
Videos
SDS