missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,5-Difluoro-N-(2,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H61127.03

Code : NEW

Additional Details : CAS Number : 931643-74-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487628

  • £53.11 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

3,5-Difluoro-N-(2,4-difluorophenyl)benzamide
269.2
18603119
931643-74-2
MFCD04806860

Specifications

3,5-Difluoro-N-(2,4-difluorophenyl)benzamide
1g
Solid
269.2
18603119
White
931643-74-2
97%
MFCD04806860
Videos
SDS