missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-2-methylbenzamide, 97%, Thermo Scientific™

Catalog Number p-7052413
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-37-5
C8H7F2NO
171.147
MFCD19687171
RASBWMSGTUCOQV-UHFFFAOYSA-N
66523572
3,5-difluoro-2-methylbenzamide
CC1=C(C=C(C=C1C(=O)N)F)F
This item is not returnable. View return policy
1323966-37-5
C8H7F2NO
171.147
MFCD19687171
RASBWMSGTUCOQV-UHFFFAOYSA-N
66523572
3,5-difluoro-2-methylbenzamide
CC1=C(C=C(C=C1C(=O)N)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 171.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Difluoro-2-methylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'