missing translation for 'onlineSavingsMsg'
Learn More

(3,5-Difluoro-2-methoxyphenyl)boronic acid, 97%, Thermo Scientific™

Catalog Number 10743983
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
737000-76-9
C7H7BF2O3
187.94
MFCD03095353
JXQHRWOUVJRCSR-UHFFFAOYSA-N
3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid
2734607
(3,5-difluoro-2-methoxyphenyl)boronic acid
COC1=C(F)C=C(F)C=C1B(O)O
This item is not returnable. View return policy
737000-76-9
C7H7BF2O3
187.94
MFCD03095353
JXQHRWOUVJRCSR-UHFFFAOYSA-N
3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid
2734607
(3,5-difluoro-2-methoxyphenyl)boronic acid
COC1=C(F)C=C(F)C=C1B(O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 200°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 187.81
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material (3, 5-Difluoro-2-methoxyphenyl)boronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (3,5-Difluoro-2-methoxyphenyl)boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'