missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-dichloropyridazine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | pyridazine, 3,5-dichloro, 3,5-dichloro-pyridazine, pyridazine,3,5-dichloro, 3,5-dichlor-pyridazin, acmc-209yx3, ksc538k8n |
Formula Weight | 148.98 |
Percent Purity | 97% |
Chemical Name or Material | 3, 5-dichloropyridazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'