missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals
39.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.
Solubility
Hydrolysis with water.
Notes
Light and moisture sensitive. Incompatible with water, alcohols, strong bases, amines, acids and strong oxidizing agents.
3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.
Solubility
Hydrolysis with water.
Notes
Light and moisture sensitive. Incompatible with water, alcohols, strong bases, amines, acids and strong oxidizing agents.
Specifications
Melting Point | 28°C to 32°C |
Density | 1.38 |
Boiling Point | 242°C to 244°C |
Flash Point | 134°C (273°F) |
Quantity | 5 g |
UN Number | UN2250 |
Beilstein | 510282 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolysis with water. |
Formula Weight | 188.01 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'